Zinc (II) phthalocyanine (Sulphate ionophore II) ≥98.0% (by titrimetric analysis), purified by sublimation
Catalog # TCIAZ0037-500MG
Supplier: TCI
CAS Number:
Specifications
- Pk:500 mg
- Seq:2725
Specifications
Specification Test Results
- Appearance:Dark red to Dark purple to Dark blue powder to crystal
- Purity (Chelometric Titration):min. 98.0 %
- Molar extinction coefficient:min. 225000 (DMF, 667 to 670nm)
Specification Test Results
About this item
:
:577.91 g/mol
EINECS:238-262-5
CAS Number:14320-04-8
Formula:C₃₂H₁₆N₈Zn
MDL Number:MFCD00041955