Zanamivir hydrate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAZ0023-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 % (calcd.on anh.substance)
- Specific rotation [a]20/D:+36.0 to +39.0 deg (C=1、H2O) (calcd.on anh.substance)
- Water:8.0 to 12.0 %
Specification Test Results
About this item
:
:332.31 g/mol (anhy)
CAS Number:551942-41-7
Formula:C₁₂H₂₀N₄O₇·xH₂O