Order Entry
Italy
ContactUsLinkComponent
Zirconium(IV) Propoxide ∼70% in 1-propanol
Flammable
Corrosive
Irritant
Health
Zirconium(IV) Propoxide ∼70% in 1-propanol
Supplier:  TCI
Certificates
Flammable
Corrosive
Irritant
Health
Zirconium(IV) Propoxide ∼70% in 1-propanol
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Yellow to Green clear liquid
  • Concentration (Chelometric Method):
    70.0 to 75.0 w/w%

Specification Test Results

Related Information

Frequently Bought Together