
WZ 811 ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:55778-02-4
:
Formula:C₁₈H₁₈N₄
MDL Number:MFCD18086914
:290.37 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:188.0 to 192.0 deg-C
Specification Test Results
Frequently Bought Together