Order Entry
Italy
ContactUsLinkComponent
Bromoethylene ∼14% ∼1 mol/L in ethyl ether
Flammable
Irritant
Health
Bromoethylene ∼14% ∼1 mol/L in ethyl ether
Catalog # TCIAV0126-100ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
Health
Bromoethylene ∼14% ∼1 mol/L in ethyl ether
Catalog # TCIAV0126-100ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Concentration (HNMR):
    13.5 to 15.0 %
  • Water:
    max. 200 ppm

Specification Test Results

About this item

 :
 :106.95 g/mol
Storage Temperature:Refrigerator
EINECS:209-800-6
UN:1993
ADR:3,I
 :GHS02
 :GHS07
 :GHS08
CAS Number:593-60-2
Formula:C₂H₃Br
MDL Number:MFCD00000183