


N-[2-(N-Vinylbenzylamino)ethyl]-3-aminopropyltrimethoxysilane hydrochloride 30 - 40% in methanol
Catalog # TCIAV0050-25ML
Supplier: TCI
CAS Number:
Specifications
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Appearance:Light yellow to Amber to Dark green clear liquid
- Concentration (with Evaporation residue):30.0 to 40.0 w/w%
Specification Test Results
About this item
:
:374.98 g/mol
EINECS:252-297-3
UN:1230
ADR:3,II
:GHS02
:GHS07
:GHS08
CAS Number:34937-00-3
Formula:C₁₇H₃₀N₂O₃Si
MDL Number:MFCD00060149