Poly(2-methyl-2-oxazoline) Amine Terminated n=∼50, ULTROXA®
Supplier: TCI
Certificates
About this item
:
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Average molecular weight:4,080 to 6,240
- Molecular weight distribution:max. 1.2
Specification Test Results
Frequently Bought Together