Poly(2-ethyl-2-oxazoline) Amine Terminated n=∼50, ULTROXA®
Catalog # TCIAU0133-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Almost white powder to lump
- Average molecular weight:4,000 to 6,000
- Molecular weight distribution:max. 1.15
Specification Test Results
About this item
:
Storage Temperature:Refrigerator
CAS Number:1616777-37-7