Order Entry
Italy
ContactUsLinkComponent
 
Poly(2-ethyl-2-oxazoline) Azide Terminated n=∼50, ULTROXA®
 :  TCI
 
Poly(2-ethyl-2-oxazoline) Azide Terminated n=∼50, ULTROXA®
 :  TCI

 

  • Appearance:
    White to Almost white powder to lump
  • Average molecular weight:
    4,000 to 6,000
  • Molecular weight distribution:
    max. 1.15

 

Frequently Bought Together