


1-[3-(Triethoxysilyl)propyl]urea 40 - 52% in methanol
Catalog # TCIAU0048-25ML
Supplier: TCI
CAS Number:
Specifications
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (with Total Nitrogen):40.0 to 52.0 w/w%
Specification Test Results
About this item
:
:264.40 g/mol
Flash Pt:14°C
EINECS:245-876-7
UN:1993
ADR:3,II
:GHS02
:GHS07
:GHS08
CAS Number:23779-32-0
Formula:C₁₀H₂₄N₂O₄Si
MDL Number:MFCD00047877