2-(p-Tolyl)-1,3-dithiane ≥98.0% (by GC)
Catalog # TCIAT3887-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:90.000 to 94.000 deg-C
Specification Test Results
About this item
:
:210.35 g/mol
CAS Number:56637-44-6
Formula:C₁₁H₁₄S₂
MDL Number:MFCD01729789