Order Entry
Italy
ContactUsLinkComponent
 
 
Tirapazamine ≥98.0% (by HPLC)
 :  TCI
 
 
Tirapazamine ≥98.0% (by HPLC)
 :  TCI

 

  • Appearance:
    Orange to Red powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Elemental analysis (Nitrogen):
    30.00 to 33.00 %

 

Frequently Bought Together