
Octadecyltrichlorosilane ≥99.0% (by GC)
TCIAT3815-1G
: TCI
:
- Pk:1 g/mol
- Seq:9220
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
:
:387.94 g/mol
:162°C
:191°C
:203-930-7
:1800
:8,II
:GHS05
:112-04-9
:C₁₈H₃₇Cl₃Si
:MFCD00000484