
2,2,6,6-Tetramethylpiperidinooxy (TEMPO) ≥99.0% (by GC, titration analysis) free radical, purified by sublimation
Supplier: TCI
Certificates
About this item
CAS Number:2564-83-2
:
Formula:C₉H₁₈NO
MDL Number:MFCD00009599
:156.25 g/mol
Merck Index:14,09140
Storage Temperature:Refrigerator
EINECS:219-888-8
UN:3263
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Orange to Brown powder to crystal
- Purity (GC):min. 99.0 %
- Purity (Iodometric Titration):min. 99.0 %
- Melting point:35.0 to 41.0 deg-C
Specification Test Results
Frequently Bought Together