Order Entry
Italy
ContactUsLinkComponent
N-[3-(Trimethoxysilyl)propyl]butan-1-amine ≥98.0% (by GC, titration analysis)
Corrosive
N-[3-(Trimethoxysilyl)propyl]butan-1-amine ≥98.0% (by GC, titration analysis)
Catalog # TCIAT3733-25ML
Supplier:  TCI
CAS Number:  
undefined
Corrosive
N-[3-(Trimethoxysilyl)propyl]butan-1-amine ≥98.0% (by GC, titration analysis)
Catalog # TCIAT3733-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :235.40 g/mol
Boiling Pt:102°C
EINECS:250-437-8
 :GHS05
CAS Number:31024-56-3
Formula:C₁₀H₂₅NO₃Si
MDL Number:MFCD02313756