2,2',7,7'-Tetrakis(diphenylamino)-9,9'-spirobi[9H-fluorene] ≥98.0% (by HPLC)
Catalog # TCIAT3634-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
Specifications
Specification Test Results
- Appearance:White to Light red to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
Specification Test Results
About this item
:
:985.25 g/mol
CAS Number:189363-47-1
Formula:C₇₃H₅₂N₄
MDL Number:MFCD11114061