
4-[2,2,2-Trifluoro-1-[(trimethylsilyl)oxy]ethyl]morpholine ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:289706-46-3
:
Formula:C₉H₁₈F₃NO₂Si
:257.33 g/mol
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together