Order Entry
Italy
ContactUsLinkComponent
Tin(II) bromide ≥97.0% (by titrimetric analysis) for perovskite precursor
Corrosive
Tin(II) bromide ≥97.0% (by titrimetric analysis) for perovskite precursor
Supplier:  TCI
Certificates
Corrosive
Tin(II) bromide ≥97.0% (by titrimetric analysis) for perovskite precursor
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (Iodometric Titration):
    min. 97.0 %

Specification Test Results

Related Information

Frequently Bought Together