
Tridodecylamine (hydrogen ionophore I) ≥85.0% (by GC)
: TCI
:102-87-4
:
:C₃₆H₇₅N
:MFCD00008971
:522 g/mol
:296°C
:190°C
:203-063-4
:GHS07
- Appearance:Colorless to Light orange to Yellow clear liquid to cloudy liquid
- Purity (GC):min. 85.0 %
- Purity (Nonaqueous Titration):min. 95.0 %
Frequently Bought Together