Order Entry
Italy
ContactUsLinkComponent
2-[2-(2,2,2-Trifluoroethoxy)phenoxy]ethyl methanesulphonate ≥98.0% (by GC)
Irritant
2-[2-(2,2,2-Trifluoroethoxy)phenoxy]ethyl methanesulphonate ≥98.0% (by GC)
Supplier:  TCI
Certificates
undefined
Irritant
2-[2-(2,2,2-Trifluoroethoxy)phenoxy]ethyl methanesulphonate ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    52.0 to 56.0 deg-C

Specification Test Results

Frequently Bought Together