Order Entry
Italy
ContactUsLinkComponent
1,4-Butanediol dimethacrylate ≥97.0% (by GC) stabilised
Irritant
1,4-Butanediol dimethacrylate ≥97.0% (by GC) stabilised
Catalog # TCIAT3488-25G
Supplier:  TCI
CAS Number:  
Irritant
1,4-Butanediol dimethacrylate ≥97.0% (by GC) stabilised
Catalog # TCIAT3488-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :226.27 g/mol
Boiling Pt:134°C
 :Stabilised with 4-methoxyphenol (MEHQ)
EINECS:218-218-1
 :GHS07
Storage Temperature:Refrigerator
CAS Number:2082-81-7
Formula:C₁₂H₁₈O₄
MDL Number:MFCD00008594