Order Entry
Italy
ContactUsLinkComponent
Tris(trimethylsilyl)phosphine ∼10% in hexane
Flammable
Irritant
Health
Tris(trimethylsilyl)phosphine ∼10% in hexane
Catalog # TCIAT3453-10G
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
Health
Tris(trimethylsilyl)phosphine ∼10% in hexane
Catalog # TCIAT3453-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Concentration (Iodometric titration):
    9.5 to 11.0 w/w%

Specification Test Results

About this item

 :
 :250.541 g/mol
Storage Temperature:Refrigerator
UN:1208
ADR:3,II
 :GHS02
 :GHS07
 :GHS08
CAS Number:15573-38-3
Formula:[(CH₃)₃Si]₃P
MDL Number:MFCD00015487