Order Entry
Italy
ContactUsLinkComponent
Trityl chloride resin 2.0-2.5 mmol/g, cross-linked with 1% DVB (200-400 mesh)
Trityl chloride resin 2.0-2.5 mmol/g, cross-linked with 1% DVB (200-400 mesh)
  TCIAT3432-1G
 :  TCI
 :  
undefined
Trityl chloride resin 2.0-2.5 mmol/g, cross-linked with 1% DVB (200-400 mesh)
  TCIAT3432-1G
 :  TCI
 :  
  • Pk:
    1 g
  • Seq:
    3047
  • Appearance:
    White to Amber powder to crystal
  • Loading:
    2.0 to 2.5 mmol/g
 :
 :GHS07
 :MFCD00284810