

2,2,2-Trifluoroethyl acetate ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:406-95-1
:
Formula:C₄H₅F₃O₂
MDL Number:MFCD00198018
:142.08 g/mol
Boiling Pt:78°C
Flash Pt:11°C
UN:3272
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together