4',6,7-Trimethoxyisoflavone ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:798-61-8
:
Formula:C₁₈H₁₆O₅
MDL Number:MFCD00016949
:312.32 g/mol
Specification Test Results
- Purity (GC):min. 97.0 %
- Melting point:177.0 to 181.0 deg-C
Specification Test Results
Frequently Bought Together