
4-(Trifluoromethyl)benzoic acid hydrazide ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:339-59-3
:
Formula:C₈H₇F₃N₂O
MDL Number:MFCD00051703
:204.15 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Bromination Method):min. 98.0 %
- Melting point:120.0 to 124.0 deg-C
Specification Test Results
Frequently Bought Together