
4-(Trifluoromethyl)styrene ≥98.0% (by GC) stabilizzato
Supplier: TCI
Certificates
About this item
CAS Number:402-50-6
:
Formula:C₉H₇F₃
MDL Number:MFCD00075539
:172.15 g/mol
Boiling Pt:66°C
Flash Pt:42°C
Storage Temperature:Refrigerata
UN:1993
ADR:3,III
:stabilizzata con 4-ter-butil pirocatecolo
:GHS02
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together