
3-(Trifluoromethyl)styrene ≥97.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:402-24-4
:
Formula:C₉H₇F₃
MDL Number:MFCD00075481
:172.15 g/mol
Boiling Pt:65°C
Flash Pt:42°C
Storage Temperature:Refrigerator
UN:1993
ADR:3,III
:Stabilised with hydroquinone (HQ)
:GHS02
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together