
Thieno[3,2-d]pyrimidin-4(1H)-one ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:16234-10-9
:MFCD03407527
Formula:C₆H₄N₂OS
MDL Number:MFCD00122140
:152.17 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:221.0 to 225.0 deg-C
Specification Test Results
Frequently Bought Together