Order Entry
Italy
ContactUsLinkComponent
5-(m-Tolyl)-5H-pyrrolo[3,2-c:4,5-c']dipyridine ≥95.0% (by GC)
Irritant
5-(m-Tolyl)-5H-pyrrolo[3,2-c:4,5-c']dipyridine ≥95.0% (by GC)
Catalog # TCIAT3281-100MG
Supplier:  TCI
CAS Number:  
Irritant
5-(m-Tolyl)-5H-pyrrolo[3,2-c:4,5-c']dipyridine ≥95.0% (by GC)
Catalog # TCIAT3281-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :259.31 g/mol
 :GHS07
CAS Number:1014403-09-8
Formula:C₁₇H₁₃N₃