Order Entry
Italy
ContactUsLinkComponent
10-(1,1':3',1''-Terphenyl-5'-yl)anthracene-9-boronic acid ≥97.0% (by HPLC) (contains varying amounts of Anhydride)
Irritant
10-(1,1':3',1''-Terphenyl-5'-yl)anthracene-9-boronic acid ≥97.0% (by HPLC) (contains varying amounts of Anhydride)
Supplier:  TCI
Certificates
undefined
Irritant
10-(1,1':3',1''-Terphenyl-5'-yl)anthracene-9-boronic acid ≥97.0% (by HPLC) (contains varying amounts of Anhydride)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Yellow powder to crystal
  • Purity (HPLC):
    min. 97.0 area%

Specification Test Results

Related Information

Frequently Bought Together