Order Entry
Italy
ContactUsLinkComponent
 
 
 
 
 
Titanium(IV) chloride ∼19% ∼1.0 mol/L in toluene
 :  TCI
 
undefined
 
 
 
 
Titanium(IV) chloride ∼19% ∼1.0 mol/L in toluene
 :  TCI

 

  • Appearance:
    Light yellow to Yellow to Orange clear liquid
  • Concentration (Argentmetric titration):
    19.0 to 22.0 w/w%

 

Frequently Bought Together