
Thieno[3,2-b]thiophene-2-carboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1723-27-9
:
Formula:C₇H₄O₂S₂
MDL Number:MFCD00115174
:184.23 g/mol
:GHS07
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together