
3,3',5,5'-Tetramethylbiphenyl-4,4'-diol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2417-04-1
:
Formula:C₁₆H₁₈O₂
MDL Number:MFCD00094737
:242.32 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:223.0 to 227.0 deg-C
Specification Test Results
Frequently Bought Together