Order Entry
Italy
ContactUsLinkComponent
 
 
3,4,5-Trimethoxybenzylamine ≥97.0% (by GC, titration analysis)
  TCIAT3087-5G
 :  TCI
 :  
undefined
 
3,4,5-Trimethoxybenzylamine ≥97.0% (by GC, titration analysis)
  TCIAT3087-5G
 :  TCI
 :  

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 97.0 %
  • Purity (Neutralization titration):
    min. 97.0 %

 

 

 :
 :197.23
 :242-468-0
 :2735
 :8,III
 :GHS05
 :18638-99-8
 :C₁₀H₁₅NO₃
 :MFCD00017150