Order Entry
Italy
ContactUsLinkComponent
2-(3-Trifluoromethylphenyl)ethylamine ≥96.0% (by GC, titration analysis)
Corrosive
2-(3-Trifluoromethylphenyl)ethylamine ≥96.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Corrosive
2-(3-Trifluoromethylphenyl)ethylamine ≥96.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green clear liquid
  • Purity (GC):
    min. 96.0 %
  • Purity (Nonaqueous Titration):
    min. 96.0 %

Specification Test Results

Related Information

Frequently Bought Together