Order Entry
Italy
ContactUsLinkComponent
1,1,2,2-Tetrafluoroethyl-2,2,3,3-tetrafluoropropyl ether ≥95.0% (by GC)
Flammable
1,1,2,2-Tetrafluoroethyl-2,2,3,3-tetrafluoropropyl ether ≥95.0% (by GC)
Supplier:  TCI
   Sustainable Options Available
Certificates
undefined
Flammable
1,1,2,2-Tetrafluoroethyl-2,2,3,3-tetrafluoropropyl ether ≥95.0% (by GC)
Supplier:  TCI
   Sustainable Options Available

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

Related Information

Frequently Bought Together