
5,6,7,7a-Tetrahydrothieno[3,2-c]pyridin-2(4H)-one hydrochloride ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:115473-15-9
:
Formula:C₇H₁₀ClNOS
MDL Number:MFCD11111130
:191.6800 g/mol
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:Light yellow to Brown to Dark green powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together