Order Entry
Italy
ContactUsLinkComponent
Thietane-1,1-dioxide ≥95.0% (by GC)
Thietane-1,1-dioxide ≥95.0% (by GC)
Catalog # TCIAT2976-200MG
Supplier:  TCI
CAS Number:  
undefined
Thietane-1,1-dioxide ≥95.0% (by GC)
Catalog # TCIAT2976-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 95.0 %
  • Melting point:
    72.0 to 76.0 deg-C

Specification Test Results

About this item

 :
 :106.14 g/mol
Boiling Pt:180°C
CAS Number:5687-92-3
Formula:C₃H₆O₂S
MDL Number:MFCD00088478