
1-(4-Methylphenyl)piperazine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:39593-08-3
:
Formula:C₁₁H₁₆N₂
MDL Number:MFCD00040737
:176.26 g/mol
Boiling Pt:112°C
EINECS:254-534-6
UN:3263
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to Orange to Green powder to lump
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together