3,3',5,5'-Tetramethylbiphenyl ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:25570-02-9
:
Formula:C₁₆H₁₈
MDL Number:MFCD00051910
:210.32 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:48.0 to 52.0 deg-C
Specification Test Results
Frequently Bought Together