4,4'-Thiodiresorcinol ≥97.0% (by GC)
Catalog # TCIAT2911-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:179.0 to 188.0 deg-C
Specification Test Results
About this item
:
EINECS:202-570-8
CAS Number:97-29-0
Formula:C₁₂H₁₀O₄S
MDL Number:MFCD00045769