
Tetrabenazine ≥99.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:58-46-8
:
Formula:C₁₉H₂₇NO₃
:317.43 g/mol
Merck Index:14,09182
EINECS:200-383-6
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (Nonaqueous Titration):min. 99.0 %
- Melting point:124.0 to 130.0 deg-C
Specification Test Results
Frequently Bought Together