
2-(Trifluoromethyl)pyridine-5-carboxaldehyde ≥98.0% (by GC)
: TCI
:386704-12-7
:
:C₇H₄F₃NO
:MFCD01862647
:175.11 g/mol
:GHS07
:Refrigerator
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:56.0 to 60.0 deg-C
Frequently Bought Together