Order Entry
Italy
ContactUsLinkComponent
Trimethyl[3-(trimethoxysilyl)propyl]ammonium chloride ∼50% in methanol
Flammable
Irritant
Health
Trimethyl[3-(trimethoxysilyl)propyl]ammonium chloride ∼50% in methanol
Supplier:  TCI
Certificates
undefined
Flammable
Irritant
Health
Trimethyl[3-(trimethoxysilyl)propyl]ammonium chloride ∼50% in methanol
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Concentration (Nonaqueous Titration):
    48.0 to 53.0 w/w%

Specification Test Results

Related Information

Frequently Bought Together