Order Entry
Italy
ContactUsLinkComponent
2,3,5,6-Tetrabromothieno[3,2-b]thiophene ≥98.0% (by GC)
2,3,5,6-Tetrabromothieno[3,2-b]thiophene ≥98.0% (by GC)
Catalog # TCIAT2772-1G
Supplier:  TCI
CAS Number:  
2,3,5,6-Tetrabromothieno[3,2-b]thiophene ≥98.0% (by GC)
Catalog # TCIAT2772-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    232.0 to 236.0 deg-C

Specification Test Results

About this item

 :
 :455.80 g/mol
CAS Number:124638-53-5
Formula:C₆Br₄S₂
MDL Number:MFCD03788229