
4,5,6,7-Tetrahydrothieno[3,2-c]pyridine hydrochloride ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:28783-41-7
:
Formula:C₇H₁₀ClNS
MDL Number:MFCD05861485
:175.68 g/mol
EINECS:249-220-0
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:226.0 to 230.0 deg-C
Specification Test Results
Frequently Bought Together