Order Entry
Italy
ContactUsLinkComponent
4,5,6,7-Tetrahydrothieno[3,2-c]pyridine ≥98.0% (by GC, titration analysis)
Irritant
4,5,6,7-Tetrahydrothieno[3,2-c]pyridine ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
4,5,6,7-Tetrahydrothieno[3,2-c]pyridine ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

Frequently Bought Together