

2-(Trifluoromethyl)pyridine ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:368-48-9
:
Formula:C₆H₄F₃N
MDL Number:MFCD02183562
:147.1 g/mol
Boiling Pt:140°C
Flash Pt:42°C
UN:1993
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together