Tetrakis[N-tetrafluorophthaloyl-(S)-tert-leucinato]dirhodium bis(ethyl acetate) adduct ≥98.0% (by HPLC)
Catalog # TCIAT2661-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Specific rotation [a]20/D:+290 to +300 deg (C=0.05, Benzene)
Specification Test Results
About this item
:
:1710.9 g/mol
CAS Number:564450-56-2
Formula:C₆₄H₅₆F₁₆N₄O₂₀Rh₂
MDL Number:MFCD15072199